missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzophenone 98.0+%, TCI America™

Catalog No. B23415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23415G Supplier TCI America Supplier No. B23415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55082-33-2
Molecular Formula C13H9BrO2
Molecular Weight (g/mol) 277.117
MDL Number MFCD00525062
InChI Key IVIICRNXAGUXLR-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxybenzophenone, 5-bromo-2-hydroxyphenyl phenyl methanone, 2-benzoyl-4-bromophenol, 5-bromo-2-hydroxyphenyl-phenylmethanone, acmc-209lkx, 5-bromo-2-hydroxy-benzophenone, 5-bromo-2-hydroxyphenyl phenyl ketone, methanone, 5-bromo-2-hydroxyphenyl phenyl, 5-bromo-2-hydroxy-phenyl-phenyl-methanone
PubChem CID 229009
IUPAC Name (5-bromo-2-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Br)O

Specifications

Melting Point 111°C
Color Yellow
Quantity 5 g
Formula Weight 277.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxybenzophenone