missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzophenone, 97%

Catalog No. p-7028282
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAB2550706 5 g
AAB2550714 25 g
2 options

Catalog No. AAB2550706

Supplier: Thermo Scientific Chemicals B2550706

Only null left

CAS: 55082-33-2 | C13H9BrO2 | 277.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55082-33-2
Molecular Formula C13H9BrO2
Molecular Weight (g/mol) 277.117
MDL Number MFCD00525062
InChI Key IVIICRNXAGUXLR-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxybenzophenone, 5-bromo-2-hydroxyphenyl phenyl methanone, 2-benzoyl-4-bromophenol, 5-bromo-2-hydroxyphenyl-phenylmethanone, acmc-209lkx, 5-bromo-2-hydroxy-benzophenone, 5-bromo-2-hydroxyphenyl phenyl ketone, methanone, 5-bromo-2-hydroxyphenyl phenyl, 5-bromo-2-hydroxy-phenyl-phenyl-methanone
PubChem CID 229009
IUPAC Name (5-bromo-2-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Br)O

Specifications

Melting Point 108°C to 111°C
Quantity 5 g
Formula Weight 277.11
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxybenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only