missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzeneboronic acid, 96%

Catalog No. p-7035223
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52937MD 250 mg
AAH5293703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52937MD Supplier Thermo Scientific Chemicals Supplier No. H52937MD

missing translation for 'validMainframeMsg'

CAS: 89598-97-0 | C6H6BBrO3 | 216.825 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89598-97-0
Molecular Formula C6H6BBrO3
Molecular Weight (g/mol) 216.825
MDL Number MFCD00093408
InChI Key XSXTWLOHAGPBNO-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxyphenyl boronic acid, 2-borono-4-bromophenol, 5-bromo-2-hydroxy benzeneboronic acid, benzeneboronic acid, 5-bromo-2-hydroxy, acmc-209r1x, 5-bromo-2-hydroxyphenyl boranediol, 5-bromo-2-hydroxybenzeneboronic acid, benzeneboronic acid,5-bromo-2-hydroxy-6ci,7ci
PubChem CID 44118764
IUPAC Name (5-bromo-2-hydroxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)Br)O)(O)O

Specifications

Melting Point 196°C to 200°C
Quantity 250 mg
Formula Weight 216.83
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only