missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine, 98%

Catalog No. p-7057327
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66885MD 250 mg
AAH6688503 1 g
AAH6688506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66885MD Supplier Thermo Scientific Chemicals Supplier No. H66885MD
Only null left

CAS: 76041-79-7 | C6H3BrF3NO | 241.995 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76041-79-7
Molecular Formula C6H3BrF3NO
Molecular Weight (g/mol) 241.995
MDL Number MFCD12546486
InChI Key OPLCXLXORZDTMX-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-trifluoromethyl pyridine, 5-bromo-3-trifluoromethyl pyridin-2-ol, 5-bromo-3-trifluoromethyl pyridin-2 1h-one, 5-bromo-3-trifluoromethyl-2 1h-pyridinone, 2 1h-pyridinone, 5-bromo-3-trifluoromethyl, 5-bromo-3-trifluoromethyl-2-pyridone, 5-bromo-3-trifluoromethyl-1h-pyridin-2-one, acmc-209p1m, ksc496m2d, 3-trifluoromethyl-5-bromo-2-pyridinol
PubChem CID 13491780
IUPAC Name 5-bromo-3-(trifluoromethyl)-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Br)C(F)(F)F

Specifications

Quantity 250 mg
Beilstein 13832576
Formula Weight 242
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only