accessibility menu, dialog, popup

UserName

CAS RN 76041-79-7

IUPAC Name: 5-bromo-3-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-3-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 242
Molecular Formula: C6H3BrF3NO
InChi Key: OPLCXLXORZDTMX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results