missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-furoic acid, 99%

Catalog No. p-7022193
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1484018 50 g
AAA1484030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1484018 Supplier Thermo Scientific Chemicals Supplier No. A1484018
Only null left

CAS: 585-70-6 | C5H3BrO3 | 190.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 585-70-6
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00003239
InChI Key YVTQHZDUDUCGRD-UHFFFAOYSA-N
Synonym 5-bromo-2-furoic acid, 5-bromofuroic acid, 5-bromo-furan-2-carboxylic acid, 5-bromo-2-furancarboxylic acid, 2-furancarboxylic acid, 5-bromo, 5-bromo-2-furanecarboxylic acid, 5-bromo furan-2-carboxylic acid, 2-furoic acid, 5-bromo, t5oj bvq ee, 5-bromofuroic?acid
PubChem CID 68511
IUPAC Name 5-bromofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)Br)C(=O)O

Specifications

Melting Point 188°C to 192°C
Quantity 50 g
Beilstein 118354
Formula Weight 190.99
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-furoic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 209-559-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only