missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-furoic Acid 98.0+%, TCI America™

$73.16 - $73.16

Chemical Identifiers

CAS 585-70-6
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00003239
InChI Key YVTQHZDUDUCGRD-UHFFFAOYSA-N
Synonym 5-bromo-2-furoic acid, 5-bromofuroic acid, 5-bromo-furan-2-carboxylic acid, 5-bromo-2-furancarboxylic acid, 2-furancarboxylic acid, 5-bromo, 5-bromo-2-furanecarboxylic acid, 5-bromo furan-2-carboxylic acid, 2-furoic acid, 5-bromo, t5oj bvq ee, 5-bromofuroic?acid
PubChem CID 68511
IUPAC Name 5-bromofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)Br)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B100825G
View Documents
TCI America
B100825G
25 g
Each for $73.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

585-70-6
190.98
YVTQHZDUDUCGRD-UHFFFAOYSA-N
68511
C1=C(OC(=C1)Br)C(=O)O
C5H3BrO3
MFCD00003239
5-bromo-2-furoic acid, 5-bromofuroic acid, 5-bromo-furan-2-carboxylic acid, 5-bromo-2-furancarboxylic acid, 2-furancarboxylic acid, 5-bromo, 5-bromo-2-furanecarboxylic acid, 5-bromo furan-2-carboxylic acid, 2-furoic acid, 5-bromo, t5oj bvq ee, 5-bromofuroic?acid
5-bromofuran-2-carboxylic acid

Specifications

585-70-6
White-Yellow
MFCD00003239
5-bromo-2-furoic acid, 5-bromofuroic acid, 5-bromo-furan-2-carboxylic acid, 5-bromo-2-furancarboxylic acid, 2-furancarboxylic acid, 5-bromo, 5-bromo-2-furanecarboxylic acid, 5-bromo furan-2-carboxylic acid, 2-furoic acid, 5-bromo, t5oj bvq ee, 5-bromofuroic?acid
C1=C(OC(=C1)Br)C(=O)O
190.98
190.98
Crystalline Powder
189°C
C5H3BrO3
25 g
YVTQHZDUDUCGRD-UHFFFAOYSA-N
5-bromofuran-2-carboxylic acid
68511
≥98.0% (HPLC,T)
5-Bromo-2-furoic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'