missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorophenol, 98%

CAS: 112204-58-7 | C6H4BrFO | 190.999 g/mol

$78.56 - $78.56

Chemical Identifiers

CAS 112204-58-7
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD07782066
InChI Key YPTHSYKJDRMAJY-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromophenol, phenol, 5-bromo-2-fluoro, 2-fluoro-5-bromo phenol, 5-bromo-2-fluoro-phenol, 3-bromo-6-fluorophenol, 3-bromo-6-fluoro-phenol, acmc-2099dd, ksc493i2d
PubChem CID 183421
IUPAC Name 5-bromo-2-fluorophenol
SMILES C1=CC(=C(C=C1Br)O)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6151806
View Documents
Thermo Scientific Chemicals
H6151806
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112204-58-7
190.999
YPTHSYKJDRMAJY-UHFFFAOYSA-N
183421
C1=CC(=C(C=C1Br)O)F
C6H4BrFO
MFCD07782066
2-fluoro-5-bromophenol, phenol, 5-bromo-2-fluoro, 2-fluoro-5-bromo phenol, 5-bromo-2-fluoro-phenol, 3-bromo-6-fluorophenol, 3-bromo-6-fluoro-phenol, acmc-2099dd, ksc493i2d
5-bromo-2-fluorophenol

Specifications

112204-58-7
53°C (0.2 mmHg)
MFCD07782066
2-fluoro-5-bromophenol, phenol, 5-bromo-2-fluoro, 2-fluoro-5-bromo phenol, 5-bromo-2-fluoro-phenol, 3-bromo-6-fluorophenol, 3-bromo-6-fluoro-phenol, acmc-2099dd, ksc493i2d
C1=CC(=C(C=C1Br)O)F
190.999
191
Crystalline
White
C6H4BrFO
5 g
YPTHSYKJDRMAJY-UHFFFAOYSA-N
5-bromo-2-fluorophenol
183421
98%
5-Bromo-2-fluorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only