missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-fluorobenzotrifluoride 98.0+%, TCI America™
$68.97 - $137.94
Chemical Identifiers
| CAS | 393-37-3 |
|---|---|
| Molecular Formula | C7H3BrF4 |
| Molecular Weight (g/mol) | 243.00 |
| MDL Number | MFCD00042493 |
| InChI Key | AJLIJYGWAXPEOK-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s |
| PubChem CID | 2736325 |
| IUPAC Name | 4-bromo-1-fluoro-2-(trifluoromethyl)benzene |
| SMILES | FC1=CC=C(Br)C=C1C(F)(F)F |
Chemical Identifiers
| 393-37-3 | |
| 243.00 | |
| AJLIJYGWAXPEOK-UHFFFAOYSA-N | |
| 2736325 | |
| FC1=CC=C(Br)C=C1C(F)(F)F |
| C7H3BrF4 | |
| MFCD00042493 | |
| 5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s | |
| 4-bromo-1-fluoro-2-(trifluoromethyl)benzene |
Specifications
| 393-37-3 | |
| 147°C | |
| MFCD00042493 | |
| 5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s | |
| FC1=CC=C(Br)C=C1C(F)(F)F | |
| 243.00 | |
| 243.00 | |
| Liquid |
| Yellow | |
| C7H3BrF4 | |
| 5 g | |
| AJLIJYGWAXPEOK-UHFFFAOYSA-N | |
| 4-bromo-1-fluoro-2-(trifluoromethyl)benzene | |
| 2736325 | |
| ≥98.0% (GC) | |
| 5-Bromo-2-fluorobenzotrifluoride |
Safety and Handling
TSCA : No