missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. B28725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28725G 5 g
B287225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28725G Supplier TCI America Supplier No. B28725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 393-37-3
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD00042493
InChI Key AJLIJYGWAXPEOK-UHFFFAOYSA-N
Synonym 5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
PubChem CID 2736325
IUPAC Name 4-bromo-1-fluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC=C(Br)C=C1C(F)(F)F

Specifications

Color Yellow
Boiling Point 147°C
Quantity 5 g
Formula Weight 243.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluorobenzotrifluoride