missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7119667
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24255G 5 g
B242525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24255G Supplier TCI America Supplier No. B24255G
Only null left

Chemical Identifiers

CAS 179897-89-3
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
InChI Key GYCNHFWRPJXTSB-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzonitrile, benzonitrile, 5-bromo-2-fluoro, 5-bromo-2-fluorobenzenecarbonitrile, 3-cyano-4-fluorobromobenzene, 3-cyano-4-fluoro-bromobenzene, 5-bromo-2-fluoro-benzonitrile, ablock ab-11-0918, pubchem1582, 2-fluoro5-bromobenzonitrile, 3-cyano-4-flurobromobenzene
PubChem CID 2724900
IUPAC Name 5-bromo-2-fluorobenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)F

Specifications

Melting Point 80°C
Quantity 5 g
UN Number 3439
Formula Weight 200.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-fluorobenzonitrile