missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzonitrile, 98%

Catalog No. AAA1611314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1611314 25 g
AAA1611306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1611314 Supplier Thermo Scientific Chemicals Supplier No. A1611314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 179897-89-3 | C7H3BrFN | 200.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 179897-89-3
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00143424
InChI Key GYCNHFWRPJXTSB-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzonitrile, benzonitrile, 5-bromo-2-fluoro, 5-bromo-2-fluorobenzenecarbonitrile, 3-cyano-4-fluorobromobenzene, 3-cyano-4-fluoro-bromobenzene, 5-bromo-2-fluoro-benzonitrile, ablock ab-11-0918, pubchem1582, 2-fluoro5-bromobenzonitrile, 3-cyano-4-flurobromobenzene
PubChem CID 2724900
IUPAC Name 5-bromo-2-fluorobenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)F

Specifications

Melting Point 77°C to 81°C
Quantity 25 g
UN Number UN3439
Beilstein 7701063
Formula Weight 200.01
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only