missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzoic Acid 97.0+%, TCI America™

Catalog No. B29741G
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B29741G 1 g
B29745G 5 g
2 options

Catalog No. B29741G

Supplier: TCI America B29741G

Only null left

Chemical Identifiers

CAS 146328-85-0
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00143423
InChI Key PEXAZYDITWXYNJ-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
PubChem CID 2736315
IUPAC Name 5-bromo-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)F

Specifications

Melting Point 150°C
Quantity 1 g
Formula Weight 219.01
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-fluorobenzoic Acid