missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-fluoroaniline 95.0+%, TCI America™
$37.29 - $143.54
Chemical Identifiers
| CAS | 2924-09-6 |
|---|---|
| Molecular Formula | C6H5BrFN |
| Molecular Weight (g/mol) | 190.02 |
| MDL Number | MFCD03094181 |
| InChI Key | ADWKOCXRCRSMLQ-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-fluorobenzenamine, 2-fluoro-5-bromoaniline, 5-bromo-2-fluoro aniline, 5-bromo-2-fluorophenylamine, benzenamine, 5-bromo-2-fluoro, 5-bromo-2-fluoro-phenylamine, 1-amino-5-bromo-2-fluorobenzene, 5-bromo-2fluoroaniline, pubchem13482, acmc-1cjje |
| PubChem CID | 2769407 |
| IUPAC Name | 5-bromo-2-fluoroaniline |
| SMILES | C1=CC(=C(C=C1Br)N)F |
Chemical Identifiers
| 2924-09-6 | |
| 190.02 | |
| ADWKOCXRCRSMLQ-UHFFFAOYSA-N | |
| 2769407 | |
| C1=CC(=C(C=C1Br)N)F |
| C6H5BrFN | |
| MFCD03094181 | |
| 5-bromo-2-fluorobenzenamine, 2-fluoro-5-bromoaniline, 5-bromo-2-fluoro aniline, 5-bromo-2-fluorophenylamine, benzenamine, 5-bromo-2-fluoro, 5-bromo-2-fluoro-phenylamine, 1-amino-5-bromo-2-fluorobenzene, 5-bromo-2fluoroaniline, pubchem13482, acmc-1cjje | |
| 5-bromo-2-fluoroaniline |
Specifications
| 2924-09-6 | |
| C6H5BrFN | |
| 5 g | |
| ADWKOCXRCRSMLQ-UHFFFAOYSA-N | |
| 5-bromo-2-fluoroaniline | |
| 2769407 | |
| ≥95.0% (GC) | |
| 5-Bromo-2-fluoroaniline |
| 105°C | |
| MFCD03094181 | |
| 5-bromo-2-fluorobenzenamine, 2-fluoro-5-bromoaniline, 5-bromo-2-fluoro aniline, 5-bromo-2-fluorophenylamine, benzenamine, 5-bromo-2-fluoro, 5-bromo-2-fluoro-phenylamine, 1-amino-5-bromo-2-fluorobenzene, 5-bromo-2fluoroaniline, pubchem13482, acmc-1cjje | |
| C1=CC(=C(C=C1Br)N)F | |
| 190.02 | |
| 190.02 | |
| Crystals |
Safety and Handling
TSCA : No