missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-fluoroaniline 95.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 105°C |
| Quantity | 5 g |
| Formula Weight | 190.02 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystals |
| Chemical Name or Material | 5-Bromo-2-fluoroaniline |
Safety and Handling
| TSCA | No |