Learn More
5-Bromo-2-fluoroaniline, 98%
CAS: 2924-09-6 | C6H5BrFN | 190.02 g/mol
$128.86 - $330.11
Chemical Identifiers
| CAS | 2924-09-6 |
|---|---|
| Molecular Formula | C6H5BrFN |
| Molecular Weight (g/mol) | 190.02 |
| MDL Number | MFCD03094181 |
| InChI Key | ADWKOCXRCRSMLQ-UHFFFAOYSA-N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6167814
|
Thermo Scientific Chemicals
H6167814 |
25 g |
Each for $128.86
|
|
|||||
|
AAH6167822
|
Thermo Scientific Chemicals
H6167822 |
100 g |
Each for $330.11
|
|
|||||
Description
5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 2924-09-6 | |
| 105°C (12 mmHg) | |
| MFCD03094181 | |
| UN2811 | |
| 2,4-diisopropylphenol, phenol, 2,4-bis 1-methylethyl, phenol, 2,4-diisopropyl, 2,4-di propan-2-yl phenol, 2,4-diisopropyl-phenol, unii-yd41xsg2l8, yd41xsg2l8, 2,4-bis 1-methylethyl phenol, propofol impurity a, acmc-20aocr | |
| ADWKOCXRCRSMLQ-UHFFFAOYSA-N | |
| 2,4-di(propan-2-yl)phenol | |
| 18048 | |
| 98% | |
| 5-Bromo-2-fluoroaniline |
| 27°C | |
| C6H5BrFN | |
| 25 g | |
| 1241528 | |
| Slightly soluble in water. | |
| NC1=C(F)C=CC(Br)=C1 | |
| 190.02 | |
| 190.01 | |
| Solid |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only