missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-2'-fluoroacetophenone, 98%

Catalog No. p-7053681
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6128206 5 g
AAH6128214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6128206 Supplier Thermo Scientific Chemicals Supplier No. H6128206

CAS: 198477-89-3 | C8H6BrFO | 217.037 g/mol

5'-Bromo-2'-fluoroacetophenone is an intermediate used in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-Bromo-2′-fluoroacetophenone is an intermediate used in pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed.

Chemical Identifiers

CAS 198477-89-3
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD11226880
InChI Key XNRQIHIOKXQSPG-UHFFFAOYSA-N
Synonym 1-5-bromo-2-fluorophenyl ethanone, 1-5-bromo-2-fluorophenyl-1-ethanone, 1-5-bromo-2-fluorophenyl ethan-1-one, ethanone, 1-5-bromo-2-fluorophenyl, 5'-bromo-2'-fluoroacetophenone, 1-acetyl-5-bromo-2-fluorobenzene, 1-5-bromo-2-fluoro-phenyl-ethanone, pubchem16430, acmc-1c1do, 5-bromo-2-fluoroacetophenone
PubChem CID 22607476
IUPAC Name 1-(5-bromo-2-fluorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Br)F

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 217.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5'-Bromo-2'-fluoroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only