missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoro-3-methylpyridine 98.0+%, TCI America™

Catalog No. B37751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37751G 1 g
B37755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37751G Supplier TCI America Supplier No. B37751G
Only null left

Chemical Identifiers

CAS 29312-98-9
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD03095091
InChI Key JLCVSFDCHKCISN-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-3-picoline, 2-fluoro-5-bromo-3-methylpyridine, pyridine, 5-bromo-2-fluoro-3-methyl, 2-fluoro-5-bromo-3-picoline, 2-fluoro-3-methyl-5-bromopyridine, 5-bromo-2-fluoro-3-methyl pyridine, pubchem1136, acmc-1cq2z, ksc495o9n, 5-bromo-2-fluoro-3-methyl-pyridine
PubChem CID 12056902
IUPAC Name 5-bromo-2-fluoro-3-methylpyridine
SMILES CC1=CC(=CN=C1F)Br

Specifications

Melting Point 65°C
Quantity 1 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-fluoro-3-methylpyridine