missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-ethoxypyridine 98.0+%, TCI America™

Catalog No. B36735G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36735G 5 g
B36731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36735G Supplier TCI America Supplier No. B36735G

Chemical Identifiers

CAS 55849-30-4
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD00234311
InChI Key WQXZKMUZWPUZGL-UHFFFAOYSA-N
Synonym 2-ethoxy-5-bromopyridine, 3-bromo-6-ethoxypyridine, 5-bromo-2-ethoxy-pyridine, pyridine, 5-bromo-2-ethoxy, zlchem 405, pubchem6235, 5-bromo-ethoxy-pyridine, acmc-1avox, 5-bromo-2-ethoxy pyridine, ksc497o6h
PubChem CID 817237
IUPAC Name 5-bromo-2-ethoxypyridine
SMILES CCOC1=NC=C(C=C1)Br

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 107°C
Quantity 5 g
Formula Weight 202.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-ethoxypyridine