accessibility menu, dialog, popup

UserName

CH3OBrN

CAS RN 55849-30-4

IUPAC Name: 5-bromo-2-ethoxypyridine
Synonyms: 5-bromo-2-ethoxypyridine
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: WQXZKMUZWPUZGL-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(Br)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results