missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyanopyrimidine, 95%

Catalog No. p-7053729
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6136303 1 g
AAH6136306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6136303 Supplier Thermo Scientific Chemicals Supplier No. H6136303
Only null left

CAS: 38275-57-9 | C5H2BrN3 | 183.996 g/mol

5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture.

Chemical Identifiers

CAS 38275-57-9
Molecular Formula C5H2BrN3
Molecular Weight (g/mol) 183.996
MDL Number MFCD02940446
InChI Key VPQICCOHFSGBMA-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyrimidine, 5-bromo-2-pyrimidinecarbonitrile, 2-pyrimidinecarbonitrile, 5-bromo, 5-bromo-pyrimidine-2-carbonitile, 2-cyano-5-bromopyrimidine, pubchem5278, acmc-1ad2b, 5-bromo-2-cyano-pyrimidine, ksc497m1f, 5-bromo-pyrimidine-2-carbonitrile
PubChem CID 686546
IUPAC Name 5-bromopyrimidine-2-carbonitrile
SMILES C1=C(C=NC(=N1)C#N)Br

Specifications

Color White to Yellow
Quantity 1 g
UN Number UN3439
Beilstein 775324
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 183.99
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-cyanopyrimidine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only