CAS RN 38275-57-9
IUPAC Name:
5-bromopyrimidine-2-carbonitrile
Synonyms:
5-bromopyrimidine-2-carbonitrile
Molecular Weight (g/mol):
184
Molecular Formula:
C5H2BrN3
InChi Key:
VPQICCOHFSGBMA-UHFFFAOYSA-N
SMILES:
BrC1=CN=C(N=C1)C#N
1
–
1
of
1
results
1
Promotions Available
5-Bromo-2-cyanopyrimidine, 95%
5-Bromo-2-cyanopyrimidine, 95%
CAS: 38275-57-9 Molecular Formula: C5H2BrN3 Molecular Weight (g/mol): 183.996 MDL Number: MFCD02940446 InChI Key: VPQICCOHFSGBMA-UHFFFAOYSA-N Synonym: 5-bromo-2-cyanopyrimidine,5-bromo-2-pyrimidinecarbonitrile,2-pyrimidinecarbonitrile, 5-bromo,5-bromo-pyrimidine-2-carbonitile,2-cyano-5-bromopyrimidine,pubchem5278,acmc-1ad2b,5-bromo-2-cyano-pyrimidine,ksc497m1f,5-bromo-pyrimidine-2-carbonitrile PubChem CID: 686546 IUPAC Name: 5-bromopyrimidine-2-carbonitrile SMILES: C1=C(C=NC(=N1)C#N)Br