missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyano-3-nitropyridine 98.0+%, TCI America™

Catalog No. p-7124124
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55845G 5 g
B558425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55845G Supplier TCI America Supplier No. B55845G

Chemical Identifiers

CAS 573675-25-9
Molecular Formula C6H2BrN3O2
Molecular Weight (g/mol) 228.005
MDL Number MFCD06657551
InChI Key OBVYONPVHIOJJZ-UHFFFAOYSA-N
Synonym 5-Bromo-3-nitropyridine-2-carbonitrile, 5-Bromo-3-nitropicolinonitrile
PubChem CID 2769698
IUPAC Name 5-bromo-3-nitropyridine-2-carbonitrile
SMILES C1=C(C(=NC=C1Br)C#N)[N+](=O)[O-]

Specifications

Melting Point 107°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 228.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-cyano-3-nitropyridine