missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorotoluene 98.0+%, TCI America™

Catalog No. p-7119885
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30025G 5 g
B300225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30025G Supplier TCI America Supplier No. B30025G
Only null left

Chemical Identifiers

CAS 54932-72-8
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00018503
InChI Key OZFQMHJKAODEON-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorotoluene, benzene, 4-bromo-1-chloro-2-methyl, 2-chloro-5-bromotoluene, pubchem3604, 5-bromo-2-chlorotoulene, acmc-209lk8, ksc493o2d, ozfqmhjkaodeon-uhfffaoysa, attercop-chm at115283
PubChem CID 609899
IUPAC Name 4-bromo-1-chloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Cl

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
Physical Form Clear Liquid at 20°C
Formula Weight 205.48
Percent Purity ≥98.0% (GC)
Chemical Name or Material 5-Bromo-2-chlorotoluene