missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorotoluene, 98%

Catalog No. p-7080870
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1060206 5 g
AAA1060214 25 g
AAA1060222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1060206 Supplier Thermo Scientific Chemicals Supplier No. A1060206

CAS: 54932-72-8 | C7H6BrCl | 205.479 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54932-72-8
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00018503
InChI Key OZFQMHJKAODEON-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorotoluene, benzene, 4-bromo-1-chloro-2-methyl, 2-chloro-5-bromotoluene, pubchem3604, 5-bromo-2-chlorotoulene, acmc-209lk8, ksc493o2d, ozfqmhjkaodeon-uhfffaoysa, attercop-chm at115283
PubChem CID 609899
IUPAC Name 4-bromo-1-chloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Cl

Specifications

Density 1.549
Quantity 5 g
Refractive Index 1.5745
Beilstein 3234896
Boiling Point 90°C to 93°C (10 mmHg)
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chlorotoluene
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only