missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloropyridine 98.0+%, TCI America™

Catalog No. B23295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23295G 5 g
B232925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23295G Supplier TCI America Supplier No. B23295G
Only null left

Chemical Identifiers

CAS 53939-30-3
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD01318951
InChI Key PEAOEIWYQVXZMB-UHFFFAOYSA-N
Synonym 2-chloro-5-bromopyridine, 5-bromo-2-chloro-pyridine, pyridine, 5-bromo-2-chloro, 2-chloro-5-bromo-pyridine, 2-chlorine-5-bromine pyridine, pubchem1168, acmc-1afhf, 3-bromo-6-chloropyridine, 2-chloro-5-bromo pyridine, 3-bromo-6-chloro-pyridine
PubChem CID 2734414
IUPAC Name 5-bromo-2-chloropyridine
SMILES C1=CC(=NC=C1Br)Cl

Specifications

Melting Point 70°C
Quantity 5 g
Formula Weight 192.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloropyridine