accessibility menu, dialog, popup

UserName

CAS RN 53939-30-3

ClBrN

CAS RN 53939-30-3

IUPAC Name: 5-bromo-2-chloropyridine
Synonyms: 5-bromo-2-chloropyridine
Molecular Weight (g/mol): 192.44
Molecular Formula: C5H3BrClN
InChi Key: PEAOEIWYQVXZMB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Br)C=N1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)