missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloropyridine, 98%

Catalog No. p-7027827
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2486206 5 g
AAB2486214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2486206 Supplier Thermo Scientific Chemicals Supplier No. B2486206
Only null left

CAS: 53939-30-3 | C5H3BrClN | 192.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53939-30-3
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD01318951
InChI Key PEAOEIWYQVXZMB-UHFFFAOYSA-N
Synonym 2-chloro-5-bromopyridine, 5-bromo-2-chloro-pyridine, pyridine, 5-bromo-2-chloro, 2-chloro-5-bromo-pyridine, 2-chlorine-5-bromine pyridine, pubchem1168, acmc-1afhf, 3-bromo-6-chloropyridine, 2-chloro-5-bromo pyridine, 3-bromo-6-chloro-pyridine
PubChem CID 2734414
IUPAC Name 5-bromo-2-chloropyridine
SMILES C1=CC(=NC=C1Br)Cl

Specifications

Melting Point 68°C to 72°C
Quantity 5 g
Beilstein 108887
Formula Weight 192.44
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chloropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only