missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorophenol 98.0+%, TCI America™

Catalog No. B29191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29191G 1 g
B29195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29191G Supplier TCI America Supplier No. B29191G

Chemical Identifiers

CAS 183802-98-4
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.451
MDL Number MFCD00672940
InChI Key UEVFFMZHGNYDKM-UHFFFAOYSA-N
Synonym 2-chloro-5-bromophenol, 5-bromo-2-chloro-phenol, phenol, 5-bromo-2-chloro, pubchem1476, acmc-1bxbv, 3-bromo-6-chlorophenol, 5-bromo-2-chloro phenol, ksc174k5h
PubChem CID 820392
IUPAC Name 5-bromo-2-chlorophenol
SMILES C1=CC(=C(C=C1Br)O)Cl

Specifications

Melting Point 58°C
Boiling Point 234°C
Quantity 1 g
Formula Weight 207.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chlorophenol