missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chlorophenol, 98+%

Catalog No. p-7028247
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2545203 1 g
AAB2545206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2545203 Supplier Thermo Scientific Chemicals Supplier No. B2545203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 183802-98-4 | C6H4BrClO | 207.451 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 183802-98-4
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.451
MDL Number MFCD00672940
InChI Key UEVFFMZHGNYDKM-UHFFFAOYSA-N
Synonym 2-chloro-5-bromophenol, 5-bromo-2-chloro-phenol, phenol, 5-bromo-2-chloro, pubchem1476, acmc-1bxbv, 3-bromo-6-chlorophenol, 5-bromo-2-chloro phenol, ksc174k5h
PubChem CID 820392
IUPAC Name 5-bromo-2-chlorophenol
SMILES C1=CC(=C(C=C1Br)O)Cl

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Formula Weight 207.46
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-chlorophenol
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only