missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloroaniline, 98%

Catalog No. p-7055504
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6410406 5 g
AAH6410414 25 g
AAH6410422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6410406 Supplier Thermo Scientific Chemicals Supplier No. H6410406

CAS: 60811-17-8 | C6H5BrClN | 206.47 g/mol

5-Bromo-2-chloroaniline is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chloroaniline is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 60811-17-8
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.47
MDL Number MFCD06796283
InChI Key UGOLEPGQWYPIBR-UHFFFAOYSA-N
Synonym 2-chloro-5-bromoaniline, 3-bromo-6-chloroaniline, 5-bromo-2-chlorophenylamine, benzenamine, 5-bromo-2-chloro, 5-bromo-2-chloro-aniline, 5-bromo-2-chloro-phenylamine, 5-bromo-2-chloro aniline, pubchem3132, 5-bromo-2-chlorobenzenamine, ksc493o4j
PubChem CID 3830731
IUPAC Name 5-bromo-2-chloroaniline
SMILES NC1=CC(Br)=CC=C1Cl

Specifications

Melting Point 145°C to 150°C
Quantity 5 g
UN Number UN2811
Beilstein 2716274
Solubility Information Slightly soluble in water.
Formula Weight 206.47
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chloroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only