missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-4-fluoroanisole 98.0+%, TCI America™

Catalog No. B5354200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5354200MG 200 mg
B53541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5354200MG Supplier TCI America Supplier No. B5354200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146447-18-9
Molecular Formula C7H5BrClFO
Molecular Weight (g/mol) 239.468
MDL Number MFCD07784265
InChI Key KHRHCBZTZQFNDK-UHFFFAOYSA-N
Synonym 1-Bromo-4-chloro-2-fluoro-5-methoxybenzene
PubChem CID 10220378
IUPAC Name 1-bromo-4-chloro-2-fluoro-5-methoxybenzene
SMILES COC1=CC(=C(C=C1Cl)F)Br

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 200 mg
Formula Weight 239.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloro-4-fluoroanisole