missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-3-nitropyridine, 98%

Catalog No. p-3735720
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379570050 5 g Glass bottle
AC379570250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC379570050 Supplier Thermo Scientific Chemicals Supplier No. 379570050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67443-38-3 | C5H2BrClN2O2 | 237.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67443-38-3
Molecular Formula C5H2BrClN2O2
Molecular Weight (g/mol) 237.44
MDL Number MFCD00222270
InChI Key WWQQPSDIIVXFOX-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-bromopyridine, 5-bromo-2-chloro-3-nitro-pyridine, pyridine, 5-bromo-2-chloro-3-nitro, abbypharma ap-12-5159, zlchem 393, pubchem1166, acmc-209nz1, ksc352q2l, 5-bromo-2-chloro-3-nitropyridin, 2-chloro-5-bromo-3-nitropyridine
PubChem CID 7019412
IUPAC Name 5-bromo-2-chloro-3-nitropyridine
SMILES [O-][N+](=O)C1=CC(Br)=CN=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 64.0°C to 69.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 237.44
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloro-3-nitropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only