accessibility menu, dialog, popup

UserName

CAS RN 67443-38-3

IUPAC Name: 5-bromo-2-chloro-3-nitropyridine
Synonyms: 5-bromo-2-chloro-3-nitropyridine
Molecular Weight (g/mol): 237.44
Molecular Formula: C5H2BrClN2O2
InChi Key: WWQQPSDIIVXFOX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CN=C1Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)