missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-chloro-3-fluoropyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 52°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 210.43 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 5-Bromo-2-chloro-3-fluoropyridine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |