CAS RN 831203-13-5
IUPAC Name:
5-bromo-2-fluoropyridin-3-amine
Synonyms:
5-bromo-2-fluoropyridin-3-amine
Molecular Weight (g/mol):
191
Molecular Formula:
C5H4BrFN2
InChi Key:
CLYOMCMDBNNGSM-UHFFFAOYSA-N
SMILES:
NC1=CC(Br)=CN=C1F
1
–
1
of
1
results
1
5-Bromo-2-chloro-3-fluoropyridine 98.0+%, TCI America™
CAS: 831203-13-5 Molecular Formula: C5H4BrFN2 Molecular Weight (g/mol): 191.00 MDL Number: MFCD03411574 InChI Key: CLYOMCMDBNNGSM-UHFFFAOYSA-N Synonym: 2-chloro-3-fluoro-5-bromopyridine,5-bromo-2-chloro-3-fluoro-pyridine,pyridine,5-bromo-2-chloro-3-fluoro,pyridine, 5-bromo-2-chloro-3-fluoro,pubchem1163,acmc-209pr9,ksc495k9j,3-bromo-6-chloro-5-fluoropyridine,3-bromo-6-chloro-5-fluoro-pyridine,5-bromo-2-chloro-3-fluoro pyridine PubChem CID: 2763285 IUPAC Name: 5-bromo-2-fluoropyridin-3-amine SMILES: NC1=CC(Br)=CN=C1F