missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-(5-bromo-2-thienyl)pyridine 98.0+%, TCI America™
$313.16 - $941.35
Chemical Identifiers
| CAS | 136902-53-9 |
|---|---|
| Molecular Formula | C9H5Br2NS |
| Molecular Weight (g/mol) | 319.014 |
| InChI Key | AZBHJODALOVOSN-UHFFFAOYSA-N |
| Synonym | 2-Bromo-5-(5-bromo-2-pyridyl)thiophene |
| PubChem CID | 12144568 |
| IUPAC Name | 5-bromo-2-(5-bromothiophen-2-yl)pyridine |
| SMILES | C1=CC(=NC=C1Br)C2=CC=C(S2)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4886200MG
|
TCI America
B4886200MG |
200 mg |
Each for $313.16
|
|
|||||
|
B48861G
|
TCI America
B48861G |
1 g |
Each for $941.35
|
|
|||||
Chemical Identifiers
| 136902-53-9 | |
| 319.014 | |
| 2-Bromo-5-(5-bromo-2-pyridyl)thiophene | |
| 5-bromo-2-(5-bromothiophen-2-yl)pyridine |
| C9H5Br2NS | |
| AZBHJODALOVOSN-UHFFFAOYSA-N | |
| 12144568 | |
| C1=CC(=NC=C1Br)C2=CC=C(S2)Br |
Specifications
| 136902-53-9 | |
| White-Yellow | |
| 200 mg | |
| AZBHJODALOVOSN-UHFFFAOYSA-N | |
| 5-bromo-2-(5-bromothiophen-2-yl)pyridine | |
| 12144568 | |
| ≥98.0% (GC) | |
| 5-Bromo-2-(5-bromo-2-thienyl)pyridine |
| 106°C | |
| C9H5Br2NS | |
| 2-Bromo-5-(5-bromo-2-pyridyl)thiophene | |
| C1=CC(=NC=C1Br)C2=CC=C(S2)Br | |
| 319.014 | |
| 319.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : No