accessibility menu, dialog, popup

UserName

NBrSBr

CAS RN 136902-53-9

IUPAC Name: 5-bromo-2-(5-bromothiophen-2-yl)pyridine
Synonyms: 5-bromo-2-(5-bromothiophen-2-yl)pyridine
Molecular Weight (g/mol): 319.01
Molecular Formula: C9H5Br2NS
InChi Key: AZBHJODALOVOSN-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=NC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results