missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethoxypyrimidine 98.0+%, TCI America™

Catalog No. B19201G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19201G 1 g
B19205G 5 g
B192025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B19201G Supplier TCI America Supplier No. B19201G
Only null left

Chemical Identifiers

CAS 56686-16-9
Molecular Formula C6H7BrN2O2
Molecular Weight (g/mol) 219.038
MDL Number MFCD00038016
InChI Key QEZIMQMMEGPYTR-UHFFFAOYSA-N
Synonym 2,4-dimethoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxy-1,3-diazine, 5-bromo-2,4-bis methyloxy pyrimidine, 5-bromo-2,4-dimethoxy-pyrimidine, pubchem5281, acmc-209lu1, ksc495s0l, 5-bromo-2,4-dimethoxypyrimidine, 5-bromo-2,4-dimethoxypyrimidine;
PubChem CID 255719
IUPAC Name 5-bromo-2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1Br)OC

Specifications

Melting Point 63°C
Color White-Yellow
Boiling Point 125°C
Quantity 1 g
Formula Weight 219.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2,4-dimethoxypyrimidine