accessibility menu, dialog, popup

UserName

H3CONBrOH3CN

CAS RN 56686-16-9

IUPAC Name: 5-bromo-2,4-dimethoxypyrimidine
Synonyms: 5-bromo-2,4-dimethoxypyrimidine
Molecular Weight (g/mol): 219.04
Molecular Formula: C6H7BrN2O2
InChi Key: QEZIMQMMEGPYTR-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C(OC)=N1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)