missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethoxypyrimidine, 97%, Thermo Scientific™

Catalog No. AC443390050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443390050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC443390050 Supplier Thermo Scientific Chemicals Supplier No. 443390050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56686-16-9
Molecular Formula C6H7BrN2O2
Molecular Weight (g/mol) 219.04
InChI Key QEZIMQMMEGPYTR-UHFFFAOYSA-N
Synonym 2,4-dimethoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxy-1,3-diazine, 5-bromo-2,4-bis methyloxy pyrimidine, 5-bromo-2,4-dimethoxy-pyrimidine, pubchem5281, acmc-209lu1, ksc495s0l, 5-bromo-2,4-dimethoxypyrimidine, 5-bromo-2,4-dimethoxypyrimidine;
PubChem CID 255719
IUPAC Name 5-bromo-2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1Br)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Boiling Point 125.0°C (17.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5g
Formula Weight 219.04
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 4-dimethoxypyrimidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only