missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7124006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51921G 1 g
B51925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51921G Supplier TCI America Supplier No. B51921G
Only null left

Chemical Identifiers

CAS 32246-20-1
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.071
MDL Number MFCD05865172
InChI Key WOPJFSQYDOHZMK-UHFFFAOYSA-N
Synonym 5-bromo-2 4-dimethoxybenzoic acid, acmc-20am5v, 5-bromo-2,4-dimethoxybenzoicacid, 2,4-dimethoxy-5-bromobenzoic acid, 2,4-dimethoxy-5-bromo-benzoic acid, 5-bromo-2,4-dimethoxy-benzoic acid, 5-bromanyl-2,4-dimethoxy-benzoic acid, benzoic acid, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxybenzoic acid
PubChem CID 13505097
IUPAC Name 5-bromo-2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1C(=O)O)Br)OC

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 1 g
Formula Weight 261.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2,4-dimethoxybenzoic Acid