missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2,4-dimethoxybenzoic acid, 97%
CAS: 32246-20-1 | C9H9BrO4 | 261.071 g/mol
Supplier: Thermo Scientific Chemicals H6059906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Bromo-2,4-dimethoxybenzoic acid | |
| 198°C to 202°C | |
| MFCD05865172 | |
| 5-bromo-2 4-dimethoxybenzoic acid, acmc-20am5v, 5-bromo-2,4-dimethoxybenzoicacid, 2,4-dimethoxy-5-bromobenzoic acid, 2,4-dimethoxy-5-bromo-benzoic acid, 5-bromo-2,4-dimethoxy-benzoic acid, 5-bromanyl-2,4-dimethoxy-benzoic acid, benzoic acid, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxybenzoic acid | |
| COC1=CC(=C(C=C1C(=O)O)Br)OC | |
| 261.071 | |
| 261.07 |
| 32246-20-1 | |
| C9H9BrO4 | |
| 5 g | |
| WOPJFSQYDOHZMK-UHFFFAOYSA-N | |
| 5-bromo-2,4-dimethoxybenzoic acid | |
| 13505097 | |
| 97% |
Chemical Identifiers
| 32246-20-1 | |
| 261.071 | |
| WOPJFSQYDOHZMK-UHFFFAOYSA-N | |
| 13505097 | |
| COC1=CC(=C(C=C1C(=O)O)Br)OC |
| C9H9BrO4 | |
| MFCD05865172 | |
| 5-bromo-2 4-dimethoxybenzoic acid, acmc-20am5v, 5-bromo-2,4-dimethoxybenzoicacid, 2,4-dimethoxy-5-bromobenzoic acid, 2,4-dimethoxy-5-bromo-benzoic acid, 5-bromo-2,4-dimethoxy-benzoic acid, 5-bromanyl-2,4-dimethoxy-benzoic acid, benzoic acid, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxybenzoic acid | |
| 5-bromo-2,4-dimethoxybenzoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only