missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethoxybenzoic acid, 97%

Catalog No. AAH6059903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6059903 1 g
AAH6059906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6059903 Supplier Thermo Scientific Chemicals Supplier No. H6059903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32246-20-1 | C9H9BrO4 | 261.071 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32246-20-1
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.071
MDL Number MFCD05865172
InChI Key WOPJFSQYDOHZMK-UHFFFAOYSA-N
Synonym 5-bromo-2 4-dimethoxybenzoic acid, acmc-20am5v, 5-bromo-2,4-dimethoxybenzoicacid, 2,4-dimethoxy-5-bromobenzoic acid, 2,4-dimethoxy-5-bromo-benzoic acid, 5-bromo-2,4-dimethoxy-benzoic acid, 5-bromanyl-2,4-dimethoxy-benzoic acid, benzoic acid, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxybenzoic acid
PubChem CID 13505097
IUPAC Name 5-bromo-2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1C(=O)O)Br)OC

Specifications

Melting Point 198°C to 202°C
Quantity 1 g
Formula Weight 261.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-2,4-dimethoxybenzoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only