Learn More
5-Bromo-2,4-dihydroxybenzoic acid, 97%
CAS: 7355-22-8 | C7H5BrO4 | 233.02 g/mol
$74.67 - $213.60
Chemical Identifiers
| CAS | 7355-22-8 |
|---|---|
| Molecular Formula | C7H5BrO4 |
| Molecular Weight (g/mol) | 233.02 |
| MDL Number | MFCD00002452 |
| InChI Key | ZRBCISXJLHZOMS-UHFFFAOYSA-N |
| Synonym | benzoic acid, 5-bromo-2,4-dihydroxy, 5-bromo-beta-resorcylic acid, 2,4-dihydroxy-5-bromobenzoic acid, 5-bromo-.beta.-resorcylic acid, .beta.-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoic acid, beta-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoicacid, 5-bromo-2,4-dihydroxybenzoicacid, 5-bromo-b-resorcylic acid |
| PubChem CID | 81814 |
| IUPAC Name | 5-bromo-2,4-dihydroxybenzoic acid |
| SMILES | OC(=O)C1=CC(Br)=C(O)C=C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1823006
|
Thermo Scientific Chemicals
A1823006 |
5 g |
Each for $74.67
|
|
|||||
|
AAA1823014
|
Thermo Scientific Chemicals
A1823014 |
25 g |
Each for $213.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7355-22-8 | |
| 233.02 | |
| ZRBCISXJLHZOMS-UHFFFAOYSA-N | |
| 81814 | |
| OC(=O)C1=CC(Br)=C(O)C=C1O |
| C7H5BrO4 | |
| MFCD00002452 | |
| benzoic acid, 5-bromo-2,4-dihydroxy, 5-bromo-beta-resorcylic acid, 2,4-dihydroxy-5-bromobenzoic acid, 5-bromo-.beta.-resorcylic acid, .beta.-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoic acid, beta-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoicacid, 5-bromo-2,4-dihydroxybenzoicacid, 5-bromo-b-resorcylic acid | |
| 5-bromo-2,4-dihydroxybenzoic acid |
Specifications
| 7355-22-8 | |
| C7H5BrO4 | |
| 5 g | |
| benzoic acid, 5-bromo-2,4-dihydroxy, 5-bromo-beta-resorcylic acid, 2,4-dihydroxy-5-bromobenzoic acid, 5-bromo-.beta.-resorcylic acid, .beta.-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoic acid, beta-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoicacid, 5-bromo-2,4-dihydroxybenzoicacid, 5-bromo-b-resorcylic acid | |
| OC(=O)C1=CC(Br)=C(O)C=C1O | |
| 233.02 | |
| 233.02 | |
| 5-Bromo-2,4-dihydroxybenzoic acid |
| ∼205°C (decomposition) | |
| MFCD00002452 | |
| 2967526 | |
| ZRBCISXJLHZOMS-UHFFFAOYSA-N | |
| 5-bromo-2,4-dihydroxybenzoic acid | |
| 81814 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-881-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only