missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dihydroxybenzoic acid, 97%

Catalog No. AAA1823014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1823014 25 g
AAA1823006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1823014 Supplier Thermo Scientific Chemicals Supplier No. A1823014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7355-22-8 | C7H5BrO4 | 233.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7355-22-8
Molecular Formula C7H5BrO4
Molecular Weight (g/mol) 233.02
MDL Number MFCD00002452
InChI Key ZRBCISXJLHZOMS-UHFFFAOYSA-N
Synonym benzoic acid, 5-bromo-2,4-dihydroxy, 5-bromo-beta-resorcylic acid, 2,4-dihydroxy-5-bromobenzoic acid, 5-bromo-.beta.-resorcylic acid, .beta.-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoic acid, beta-resorcylic acid, 5-bromo, 5-bromo-2,4-dihydroxy-benzoicacid, 5-bromo-2,4-dihydroxybenzoicacid, 5-bromo-b-resorcylic acid
PubChem CID 81814
IUPAC Name 5-bromo-2,4-dihydroxybenzoic acid
SMILES OC(=O)C1=CC(Br)=C(O)C=C1O

Specifications

Melting Point ∼205°C (decomposition)
Quantity 25 g
Beilstein 2967526
Formula Weight 233.02
Percent Purity 98%
Chemical Name or Material 5-Bromo-2,4-dihydroxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-881-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only