missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydro-1-benzofuran, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC00810EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC00810EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

66826-78-6
199.05
UDWFSJAYXTXMLM-UHFFFAOYSA-N
2776159
BrC1=CC2=C(OCC2)C=C1
C8H7BrO
MFCD02677716
5-bromo-2,3-dihydrobenzofuran, 5-bromo-2,3-dihydrobenzo b furan, 5-bromocoumaran, benzofuran, 5-bromo-2,3-dihydro, 2,3-dihydro-5-bromobenzofuran, buttpark 36\18-16, pubchem7055, 5-bromo-2,3 dihydrobenzofuran, acmc-1b981, 5-bromo-2,3-dihydro-benzofuran
5-bromo-2,3-dihydro-1-benzofuran

Specifications

5-Bromo-2, 3-dihydro-1-benzofuran
96.5
97%
C8H7BrO
MFCD02677716
UDWFSJAYXTXMLM-UHFFFAOYSA-N
5-bromo-2,3-dihydro-1-benzofuran
2776159
97%
66826-78-6
100.0
Amber Glass Bottle
10g
5-bromo-2,3-dihydrobenzofuran, 5-bromo-2,3-dihydrobenzo b furan, 5-bromocoumaran, benzofuran, 5-bromo-2,3-dihydro, 2,3-dihydro-5-bromobenzofuran, buttpark 36\18-16, pubchem7055, 5-bromo-2,3 dihydrobenzofuran, acmc-1b981, 5-bromo-2,3-dihydro-benzofuran
BrC1=CC2=C(OCC2)C=C1
199.05
199.05
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions