accessibility menu, dialog, popup

UserName

OBr

CAS RN 66826-78-6

IUPAC Name: 5-bromo-2,3-dihydro-1-benzofuran
Synonyms: 5-bromo-2,3-dihydro-1-benzofuran
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: UDWFSJAYXTXMLM-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCC2)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)