missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-difluoropyridine, 97%

Catalog No. AAH6456614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6456614 25 g
AAH6456603 1 g
AAH6456606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6456614 Supplier Thermo Scientific Chemicals Supplier No. H6456614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89402-44-8 | C5H2BrF2N | 193.98 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 89402-44-8
Molecular Formula C5H2BrF2N
Molecular Weight (g/mol) 193.98
MDL Number MFCD06659525
InChI Key QVIQXJRQVOPYGI-UHFFFAOYSA-N
PubChem CID 14549654
IUPAC Name 5-bromo-2,3-difluoropyridine
SMILES FC1=CC(Br)=CN=C1F

Specifications

Color Colorless to Yellow
Quantity 25 g
Beilstein 13412561
Synonym pyridine, 5-bromo-2,3-difluoro, 2,3-difluoro-5-bromopyridine, pubchem1292, 5-bromo-2,3-difluoro-pyridine, pyridine,5-bromo-2,3-difluoro, 5-bromanyl-2,3-bis fluoranyl pyridine
Solubility Information Slightly soluble in water.
Formula Weight 193.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2,3-difluoropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only