accessibility menu, dialog, popup

UserName

CAS RN 89402-44-8

NBrFF

CAS RN 89402-44-8

IUPAC Name: 5-bromo-2,3-difluoropyridine
Synonyms: 5-bromo-2,3-difluoropyridine
Molecular Weight (g/mol): 193.98
Molecular Formula: C5H2BrF2N
InChi Key: QVIQXJRQVOPYGI-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CN=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results